Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Xt03687300_m1
          See other TNNT3 GE Assays ›
          Gene Symbol:
          tnnt3
          Gene Name:
          troponin T3, fast skeletal type
          Gene Aliases:
          amcd2b, da2b, fssv
          Chromosome Location:
          Chr.4: 12626550 - 12647760 on Build UCB_Xtro_10.0
          Species:
          Xenopus tropicalis
          Species Specific ID (Flybase ID):
          -
          Assay ID Xt03687300_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: tnnt3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_203812.1 NP_989143.1 7 - 8 269 75
          XM_012960736.2 XP_012816190.1 11 - 12 474 75
          XM_012960737.2 XP_012816191.1 11 - 12 474 75
          XM_012960738.2 XP_012816192.1 10 - 11 462 75
          XM_012960739.2 XP_012816193.1 10 - 11 462 75
          XM_012960741.2 XP_012816195.1 10 - 11 453 75
          XM_012960742.2 XP_012816196.1 10 - 11 453 75
          XM_012960743.2 XP_012816197.1 9 - 10 441 75
          XM_012960744.2 XP_012816198.1 9 - 10 441 75
          XM_012960745.2 XP_012816199.1 10 - 11 408 75
          XM_012960746.2 XP_012816200.1 10 - 11 408 75
          XM_012960748.2 XP_012816202.1 9 - 10 396 75
          XM_012960750.2 XP_012816204.1 9 - 10 387 75
          XM_012960751.2 XP_012816205.1 9 - 10 387 75
          XM_012960752.2 XP_012816206.1 8 - 9 375 75
          XM_012960753.2 XP_012816207.1 8 - 9 375 75
          XM_012960754.2 XP_012816208.1 8 - 9 372 75
          XM_012960755.2 XP_012816209.1 8 - 9 372 75
          XM_012960756.2 XP_012816210.1 6 - 7 342 75
          XM_012960758.2 XP_012816212.1 6 - 7 342 75
          XM_018092959.1 XP_017948448.1 11 - 12 474 75
          XM_018092960.1 XP_017948449.1 10 - 11 462 75
          XM_018092961.1 XP_017948450.1 10 - 11 459 75
          XM_018092962.1 XP_017948451.1 10 - 11 453 75
          XM_018092963.1 XP_017948452.1 9 - 10 441 75
          XM_018092964.1 XP_017948453.1 10 - 11 408 75
          XM_018092965.1 XP_017948454.1 9 - 10 396 75
          XM_018092966.1 XP_017948455.1 9 - 10 393 75
          XM_018092967.1 XP_017948456.1 9 - 10 387 75
          XM_018092968.1 XP_017948457.1 8 - 9 375 75
          XM_018092969.1 XP_017948458.1 8 - 9 372 75
          XM_018092970.1 XP_017948459.1 6 - 7 342 75
          GenBank mRNA BC061295.1 - 7 - 8 269 75
          CR760479.2 - 10 - 11 393 75

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons

          Panther Classification:

          Molecular Function -

          actin binding motor protein

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline