Search
Search

ubqln1| Gene Symbol: ubqln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203890.1 | NP_989221.1 | 7 - 8 | 1447 | 79 | |
| GenBank mRNA | BC063190.1 | - | 7 - 8 | 1447 | 79 | |
| CT025388.2 | - | 7 - 8 | 1409 | 79 | ||
Set Membership: |
|