Search
Search

rabgef1| Gene Symbol: rabgef1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204000.1 | NP_989331.1 | 5 - 6 | 656 | 79 | |
| XM_012956676.2 | XP_012812130.1 | 5 - 6 | 711 | 79 | ||
| XM_012956677.2 | XP_012812131.1 | 5 - 6 | 728 | 79 | ||
| XM_012956679.2 | XP_012812133.1 | 5 - 6 | 619 | 79 | ||
| GenBank mRNA | BC064156.1 | - | 5 - 6 | 656 | 79 | |
Set Membership: |
|