Search
Search

rcn3| Gene Symbol: rcn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204058.1 | NP_989389.1 | 3 - 4 | 447 | 89 | |
| GenBank mRNA | BC064883.1 | - | 3 - 4 | 447 | 89 | |
| CT030617.1 | - | 3 - 4 | 446 | 89 | ||
Set Membership: |
|