Search
Search

actb| Gene Symbol: actb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213719.1 | NP_998884.1 | 3 - 4 | 394 | 73 | |
| GenBank mRNA | BC068217.1 | - | 3 - 4 | 394 | 73 | |
| BC082343.1 | - | 3 - 4 | 395 | 73 | ||
| CR855434.2 | - | 2 - 3 | 388 | 73 | ||
Set Membership: |
|