Search
Search

odc1| Gene Symbol: odc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005441.1 | NP_001005441.1 | 7 - 8 | 977 | 76 | |
| GenBank mRNA | BC074547.1 | - | 7 - 8 | 977 | 76 | |
| CT025275.2 | - | 5 - 6 | 672 | 76 | ||
Set Membership: |
|