Search
Search

dpysl3| Gene Symbol: dpysl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005637.1 | NP_001005637.1 | 3 - 4 | 363 | 97 | |
| XM_012958802.2 | XP_012814256.1 | 3 - 4 | 639 | 97 | ||
| XM_012958803.2 | XP_012814257.1 | 3 - 4 | 639 | 97 | ||
| GenBank mRNA | BC074633.1 | - | 3 - 4 | 363 | 97 | |
| CR942629.1 | - | 3 - 4 | 364 | 97 | ||
Set Membership: |
|