Search
Search

papolg| Gene Symbol: papolg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005684.1 | NP_001005684.1 | 16 - 17 | 1699 | 91 | |
| XM_012962875.2 | XP_012818329.1 | 15 - 16 | 1725 | 91 | ||
| GenBank mRNA | BC075107.1 | - | 16 - 17 | 1699 | 91 | |
Set Membership: |
|