Search
Search

pcolce2| Gene Symbol: pcolce2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005804.1 | NP_001005804.1 | 6 - 7 | 999 | 72 | |
| GenBank mRNA | BC075353.1 | - | 6 - 7 | 999 | 72 | |
| CR760811.2 | - | 6 - 7 | 953 | 72 | ||
Set Membership: |
|