Search
Search

atf1| Gene Symbol: atf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005828.2 | NP_001005828.1 | 6 - 7 | 763 | 64 | |
| GenBank mRNA | BC075378.1 | - | 6 - 7 | 765 | 64 | |
| CT030449.1 | - | 6 - 7 | 767 | 64 | ||
| CU075764.1 | - | 6 - 7 | 771 | 64 | ||
Set Membership: |
|