Search
Search

spa17| Gene Symbol: spa17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006757.1 | NP_001006758.1 | 5 - 6 | 560 | 102 | |
| XM_012966399.2 | XP_012821853.1 | 5 - 6 | 831 | 102 | ||
| XM_012966400.2 | XP_012821854.1 | 5 - 6 | 946 | 102 | ||
| XM_012966402.2 | XP_012821856.1 | 4 - 5 | 709 | 102 | ||
| GenBank mRNA | BC075539.1 | - | 5 - 6 | 560 | 102 | |
| CT030312.1 | - | 610 | 102 | |||
Set Membership: |
|