Search
Search

ncbp1| Gene Symbol: ncbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006787.1 | NP_001006788.1 | 16 - 17 | 1698 | 199 | |
| GenBank mRNA | BC075600.1 | - | 16 - 17 | 1698 | 199 | |
| CR855671.2 | - | 16 - 17 | 1725 | 199 | ||
Set Membership: |
|