Search
Search

ctdsp2| Gene Symbol: ctdsp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006792.1 | NP_001006793.1 | 2 - 3 | 607 | 96 | |
| XM_012956751.2 | XP_012812205.1 | 3 - 4 | 1069 | 96 | ||
| GenBank mRNA | BC076658.1 | - | 2 - 3 | 607 | 96 | |
Set Membership: |
|