Search
Search

cdk18| Gene Symbol: cdk18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006836.1 | NP_001006837.1 | 12 - 13 | 1618 | 79 | |
| GenBank mRNA | BC076915.1 | - | 12 - 13 | 1618 | 79 | |
Set Membership: |
|