Search
Search

mcts1| Gene Symbol: mcts1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006879.1 | NP_001006880.1 | 4 - 5 | 543 | 88 | |
| GenBank mRNA | BC077034.1 | - | 4 - 5 | 543 | 88 | |
| CR761407.2 | - | 4 - 5 | 454 | 88 | ||
Set Membership: |
|