Search
Search

ephx2| Gene Symbol: ephx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006911.1 | NP_001006912.1 | 16 - 17 | 1493 | 66 | |
| XM_012964034.2 | XP_012819488.1 | 16 - 17 | 1630 | 66 | ||
| GenBank mRNA | BC075370.1 | - | 16 - 17 | 1493 | 66 | |
Set Membership: |
|