Search
Search

kras| Gene Symbol: kras | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008033.1 | NP_001008034.1 | 4 - 5 | 655 | 84 | |
| GenBank mRNA | BC080916.1 | - | 4 - 5 | 655 | 84 | |
| CR760764.2 | - | 4 - 5 | 655 | 84 | ||
Set Membership: |
|