Search
Search

ctnnbip1| Gene Symbol: ctnnbip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008102.1 | NP_001008103.1 | 3 - 4 | 283 | 82 | |
| GenBank mRNA | BC081305.1 | - | 3 - 4 | 283 | 82 | |
| CR855834.2 | - | 3 - 4 | 303 | 82 | ||
Set Membership: |
|