Search
Search

nup35| Gene Symbol: nup35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008123.1 | NP_001008124.1 | 2 - 3 | 271 | 89 | |
| XM_012970344.2 | XP_012825798.1 | 3 | 107 | |||
| XM_012970345.2 | XP_012825799.1 | 3 | 107 | |||
| XM_012970346.1 | XP_012825800.1 | 2 - 3 | 302 | 89 | ||
| GenBank mRNA | BC081329.1 | - | 2 - 3 | 271 | 89 | |
| CR761020.2 | - | 2 - 3 | 204 | 89 | ||
Set Membership: |
|