Search
Search

pcolce| Gene Symbol: pcolce | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008156.1 | NP_001008157.1 | 5 - 6 | 1161 | 86 | |
| GenBank mRNA | BC081370.1 | - | 5 - 6 | 1161 | 86 | |
| BC090811.1 | - | 5 - 6 | 1031 | 86 | ||
Set Membership: |
|