Search
Search

ctbp2| Gene Symbol: ctbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012967715.2 | XP_012823169.1 | 9 - 10 | 3250 | 61 | |
| GenBank mRNA | BC082524.1 | - | 8 - 9 | 1354 | 61 | |
| CR760305.2 | - | 5 - 6 | 513 | 61 | ||
Set Membership: |
|