Search
Search

trub2| Gene Symbol: trub2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011086.1 | NP_001011086.1 | 282 | 85 | ||
| GenBank mRNA | BC084478.1 | - | 282 | 85 | ||
| CR761544.2 | - | 220 | 85 | |||
Set Membership: |
|