Search
Search

lap3| Gene Symbol: lap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011124.2 | NP_001011124.1 | 10 - 11 | 1198 | 62 | |
| XM_012960775.2 | XP_012816229.1 | 10 - 11 | 1135 | 62 | ||
| XM_018092770.1 | XP_017948259.1 | 10 - 11 | 1274 | 62 | ||
| XM_018092777.1 | XP_017948266.1 | 10 - 11 | 1132 | 62 | ||
| GenBank mRNA | BC084523.1 | - | 10 - 11 | 1200 | 62 | |
| CT027972.2 | - | 10 - 11 | 1209 | 62 | ||
Set Membership: |
|