Search
Search

esd| Gene Symbol: esd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011247.1 | NP_001011247.1 | 3 - 4 | 228 | 99 | |
| XM_012956849.2 | XP_012812303.1 | 2 - 3 | 782 | 99 | ||
| XM_018091202.1 | XP_017946691.1 | 3 - 4 | 973 | 99 | ||
| GenBank mRNA | BC087823.1 | - | 3 - 4 | 228 | 99 | |
| CT030509.1 | - | 3 - 4 | 191 | 99 | ||
Set Membership: |
|