Search
Search

serpinb5| Gene Symbol: serpinb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011282.1 | NP_001011282.1 | 3 - 4 | 365 | 107 | |
| GenBank mRNA | BC087996.1 | - | 3 - 4 | 365 | 107 | |
Set Membership: |
|