Search
Search

emc10| Gene Symbol: emc10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203571.1 | NP_988902.1 | 2 - 3 | 198 | 90 | |
| XM_012956621.2 | XP_012812075.1 | 2 - 3 | 285 | 90 | ||
| GenBank mRNA | BC061625.1 | - | 2 - 3 | 198 | 90 | |
| CR760224.2 | - | 2 - 3 | 203 | 90 | ||
Set Membership: |
|