Search
Search

rspo2| Gene Symbol: rspo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011386.1 | NP_001011386.1 | 2 - 3 | 405 | 73 | |
| XM_012964623.2 | XP_012820077.1 | 3 - 4 | 1770 | 73 | ||
| GenBank mRNA | BC088569.1 | - | 2 - 3 | 405 | 73 | |
Set Membership: |
|