Search
Search

asmt| Gene Symbol: asmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011409.1 | NP_001011409.1 | 4 - 5 | 401 | 85 | |
| XM_012956873.2 | XP_012812327.1 | 4 - 5 | 513 | 85 | ||
| GenBank mRNA | BC088783.1 | - | 4 - 5 | 401 | 85 | |
Set Membership: |
|