Search
Search

sike1| Gene Symbol: sike1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011474.1 | NP_001011474.1 | 2 - 3 | 287 | 97 | |
| XM_012961449.2 | XP_012816903.1 | 2 - 3 | 2010 | 97 | ||
| XM_012961451.1 | XP_012816905.1 | 3 - 4 | 479 | 97 | ||
| GenBank mRNA | BC088764.1 | - | 2 - 3 | 287 | 97 | |
| CR848255.2 | - | 2 - 3 | 285 | 97 | ||
Set Membership: |
|