Search
Search

xk| Gene Symbol: xk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012289.1 | NP_001012289.1 | 1 - 2 | 329 | 90 | |
| GenBank mRNA | AY534993.1 | - | 1 - 2 | 329 | 90 | |
| BC171200.1 | - | 1 - 2 | 319 | 90 | ||
| BC171204.1 | - | 1 - 2 | 320 | 90 | ||
Set Membership: |
|