Search
Search

bnip3| Gene Symbol: bnip3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015859.1 | NP_001015859.1 | 4 - 5 | 498 | 58 | |
| XM_012966550.2 | XP_012822004.1 | 4 - 5 | 639 | 58 | ||
| XM_018095382.1 | XP_017950871.1 | 4 - 5 | 647 | 58 | ||
| XM_018095383.1 | XP_017950872.1 | 4 - 5 | 642 | 58 | ||
| GenBank mRNA | BC090370.1 | - | 4 - 5 | 498 | 58 | |
| CT027850.1 | - | 4 - 5 | 480 | 58 | ||
Set Membership: |
|