Search
Search

trappc3| Gene Symbol: trappc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015966.2 | NP_001015966.1 | 4 - 5 | 477 | 104 | |
| GenBank mRNA | BC091013.1 | - | 4 - 5 | 499 | 104 | |
| CR848424.2 | - | 4 - 5 | 477 | 104 | ||
Set Membership: |
|