Search
Search

lsm2| Gene Symbol: lsm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016402.3 | NP_001016402.1 | 2 - 3 | 208 | 80 | |
| GenBank mRNA | CN108372.1 | - | 2 - 3 | 210 | 80 | |
| CR761142.1 | - | 2 - 3 | 87 | 80 | ||
| DN095519.1 | - | 2 - 3 | 131 | 80 | ||
| DR900519.1 | - | 2 - 3 | 180 | 80 | ||
Set Membership: |
|