Search
Search

camkmt| Gene Symbol: camkmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016430.3 | NP_001016430.2 | 1 - 2 | 197 | 57 | |
| GenBank mRNA | BC171086.1 | - | 1 - 2 | 124 | 57 | |
| CN090931.1 | - | 1 - 2 | 196 | 57 | ||
| CR761052.2 | - | 1 - 2 | 190 | 57 | ||
| CX886540.1 | - | 1 - 2 | 197 | 57 | ||
Set Membership: |
|