Search
Search

ucma| Gene Symbol: ucma | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016485.2 | NP_001016485.1 | 4 - 5 | 319 | 104 | |
| GenBank mRNA | BC135882.1 | - | 4 - 5 | 333 | 104 | |
| CR760848.2 | - | 4 - 5 | 319 | 104 | ||
Set Membership: |
|