Search
Search

frzb2| Gene Symbol: frzb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012961122.2 | XP_012816576.1 | 2 - 3 | 700 | 68 | |
| GenBank mRNA | BC170567.1 | - | 2 - 3 | 431 | 68 | |
| BC170569.1 | - | 2 - 3 | 431 | 68 | ||
| CR760192.1 | - | 2 - 3 | 531 | 68 | ||
Set Membership: |
|