Search
Search

vkorc1l1| Gene Symbol: vkorc1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016769.3 | NP_001016769.2 | 1 - 2 | 196 | 103 | |
| GenBank mRNA | BC166389.1 | - | 1 - 2 | 189 | 103 | |
| CR855740.2 | - | 1 - 2 | 196 | 103 | ||
Set Membership: |
|