Search
Search

orc5| Gene Symbol: orc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017013.2 | NP_001017013.1 | 8 - 9 | 1015 | 74 | |
| XM_012964762.2 | XP_012820216.1 | 9 - 10 | 1000 | 74 | ||
| XM_012964764.2 | XP_012820218.1 | 8 - 9 | 1093 | 74 | ||
| GenBank mRNA | BC136000.1 | - | 8 - 9 | 3740 | 74 | |
| CR762239.2 | - | 8 - 9 | 1015 | 74 | ||
Set Membership: |
|