Search
Search

ndufb10| Gene Symbol: ndufb10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017250.2 | NP_001017250.1 | 3 - 4 | 442 | 71 | |
| GenBank mRNA | BC135549.1 | - | 3 - 4 | 447 | 71 | |
| CR760102.2 | - | 3 - 4 | 442 | 71 | ||
Set Membership: |
|