Search
Search

slc20a1| Gene Symbol: slc20a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030397.1 | NP_001025568.1 | 5 - 6 | 1021 | 94 | |
| XM_012959125.2 | XP_012814579.1 | 5 - 6 | 1062 | 94 | ||
| GenBank mRNA | BC090608.1 | - | 5 - 6 | 1021 | 94 | |
Set Membership: |
|