Search
Search

matn1| Gene Symbol: matn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030442.1 | NP_001025613.1 | 3 - 4 | 672 | 82 | |
| XM_018091297.1 | XP_017946786.1 | 3 - 4 | 705 | 82 | ||
| GenBank mRNA | BC091071.1 | - | 3 - 4 | 672 | 82 | |
Set Membership: |
|