Search
Search

cdv3| Gene Symbol: cdv3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004958.2 | NP_001004958.1 | 2 - 3 | 362 | 82 | |
| XM_018094615.1 | XP_017950104.1 | 2 - 3 | 597 | 82 | ||
| GenBank mRNA | BC075463.1 | - | 2 - 3 | 368 | 82 | |
| CR926414.2 | - | 2 - 3 | 362 | 82 | ||
Set Membership: |
|