Search
Search

cdc16| Gene Symbol: cdc16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005030.2 | NP_001005030.1 | 9 - 10 | 1133 | 107 | |
| GenBank mRNA | BC076877.1 | - | 9 - 10 | 1052 | 107 | |
| CR762096.2 | - | 9 - 10 | 1133 | 107 | ||
Set Membership: |
|