Search
Search

hey1| Gene Symbol: hey1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007910.2 | NP_001007911.1 | 2 - 3 | 327 | 103 | |
| GenBank mRNA | BC080349.1 | - | 2 - 3 | 287 | 103 | |
| CR760684.2 | - | 2 - 3 | 327 | 103 | ||
Set Membership: |
|