Search
Search

c7orf26| Gene Symbol: c7orf26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016231.2 | NP_001016231.1 | 3 - 4 | 459 | 74 | |
| GenBank mRNA | BC123054.1 | - | 1 - 2 | 99 | 74 | |
| BC171225.1 | - | 3 - 4 | 459 | 74 | ||
| BC171227.1 | - | 3 - 4 | 459 | 74 | ||
| CR926283.2 | - | 3 - 4 | 459 | 74 | ||
Set Membership: |
|