Search
Search

mzt1| Gene Symbol: mzt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016381.2 | NP_001016381.1 | 2 - 3 | 200 | 65 | |
| GenBank mRNA | BC118868.1 | - | 2 - 3 | 65 | ||
| CR761220.2 | - | 2 - 3 | 200 | 65 | ||
Set Membership: |
|