Search
Search

eloc| Gene Symbol: eloc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016420.2 | NP_001016420.1 | 2 - 3 | 205 | 94 | |
| XM_012964699.2 | XP_012820153.1 | 3 - 4 | 340 | 94 | ||
| XM_012964700.2 | XP_012820154.1 | 3 - 4 | 496 | 94 | ||
| XM_018094652.1 | XP_017950141.1 | 3 - 4 | 344 | 94 | ||
| GenBank mRNA | BC135463.1 | - | 3 - 4 | 237 | 94 | |
| CR761092.2 | - | 2 - 3 | 205 | 94 | ||
Set Membership: |
|