Search
Search

lman1l| Gene Symbol: lman1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016497.2 | NP_001016497.1 | 2 - 3 | 515 | 86 | |
| GenBank mRNA | BC121218.1 | - | 2 - 3 | 585 | 86 | |
| CR760808.2 | - | 2 - 3 | 515 | 86 | ||
Set Membership: |
|