Search
Search

cotl1| Gene Symbol: cotl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016541.2 | NP_001016541.1 | 2 - 3 | 260 | 88 | |
| GenBank mRNA | BC135722.1 | - | 2 - 3 | 322 | 88 | |
| CR760762.2 | - | 2 - 3 | 260 | 88 | ||
Set Membership: |
|