Search
Search

aurka| Gene Symbol: aurka | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016570.2 | NP_001016570.1 | 962 | 100 | ||
| GenBank mRNA | BC169141.1 | - | 1011 | 100 | ||
| CR760668.2 | - | 962 | 100 | |||
Set Membership: |
|